Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%
Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%
Catalog # APOSOR32351-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%
Catalog # APOSOR32351-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.711 g/mol
Storage Temperature:Ambient
CAS Number:37801-47-1
Formula:C₁₂H₁₄ClN₃O₂
MDL Number:MFCD04124526