6-(4-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
Catalog # APOSOR32337-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:258.706 g/mol
Storage Temperature:Ambient
CAS Number:85841-04-9
Formula:C₁₃H₁₁ClN₄
MDL Number:MFCD02661637