Order Entry
Czech Republic
ContactUsLinkComponent
5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine, tech
5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine, tech
Catalog # APOSOR32329-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine, tech
Catalog # APOSOR32329-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.288 g/mol
Storage Temperature:Ambient
CAS Number:477863-87-9
Formula:C₁₄H₁₅N₃O
MDL Number:MFCD02102339