Order Entry
Czech Republic
ContactUsLinkComponent
5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine, tech
5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine, tech
Catalog # APOSOR32328-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine, tech
Catalog # APOSOR32328-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.251 g/mol
Storage Temperature:Ambient
CAS Number:477863-75-5
Formula:C₁₂H₁₃N₃O
MDL Number:MFCD02102335