3-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)butan-2-one, tech
Catalog # APOSOR32324-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:296.947 g/mol
Storage Temperature:Ambient
CAS Number:320424-38-2
Formula:C₆H₇Br₂N₃O
MDL Number:MFCD01568885