4-Acetyl-4-pentylcyclohexan-1-one, tech
Catalog # APOSOR32321-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.313 g/mol
Storage Temperature:Ambient
CAS Number:76921-65-8
Formula:C₁₃H₂₂O₂
MDL Number:MFCD02082370