4-((Dimethylamino)methyl)benzonitrile
Catalog # APOSOR323176-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:160.216 g/mol
Storage Temperature:Ambient
CAS Number:35525-86-1
Formula:C₁₀H₁₂N₂
MDL Number:MFCD00029578