3-(2,6-Dimethylphenoxy)butan-2-one, tech
Catalog # APOSOR32313-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.254 g/mol
Storage Temperature:Ambient
CAS Number:25826-38-4
Formula:C₁₂H₁₆O₂
MDL Number:MFCD00243586