Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3,5-Dimethylphenoxy)butan-2-one
3-(3,5-Dimethylphenoxy)butan-2-one, tech
3-(3,5-Dimethylphenoxy)butan-2-one, tech
APOSOR32307-1G
:
Apollo Scientific
:
3-(3,5-Dimethylphenoxy)butan-2-one, tech
APOSOR32307-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.254 g/mol
:
Ambient
:
24264-52-6
:
C₁₂H₁₆O₂
:
MFCD00243575