3-(3-Methylphenoxy)butan-2-one, tech
Catalog # APOSOR32306-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.228 g/mol
Storage Temperature:Ambient
CAS Number:6560-02-7
Formula:C₁₁H₁₄O₂
MDL Number:MFCD00243573