3-(3-Methoxyphenoxy)butan-2-one, tech
Catalog # APOSOR32305-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.227 g/mol
Storage Temperature:Ambient
CAS Number:93351-44-1
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00243572