3-(2-Chlorophenoxy)butan-2-one, tech
Catalog # APOSOR32304-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.646 g/mol
Storage Temperature:Ambient
CAS Number:3757-62-8
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD00243571