Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,2-Dichloroacetyl)pyrazolidin-3-one, tech
1-(2,2-Dichloroacetyl)pyrazolidin-3-one, tech
Catalog # APOSOR32282-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Dichloroacetyl)pyrazolidin-3-one, tech
Catalog # APOSOR32282-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.019 g/mol
Storage Temperature:Ambient
CAS Number:303994-70-9
Formula:C₅H₆Cl₂N₂O₂
MDL Number:MFCD00127150