Order Entry
Czech Republic
ContactUsLinkComponent
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
Catalog # APOSOR322688-100MG
Supplier:  Apollo Scientific
CAS Number:  
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
Catalog # APOSOR322688-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :267.281 g/mol
Storage Temperature:Ambient
CAS Number:518990-56-2
Formula:C₁₂H₁₇N₃O₄
MDL Number:MFCD10697132