Order Entry
Czech Republic
ContactUsLinkComponent
cis-4-Cyclopentene-1,3-diol
cis-4-Cyclopentene-1,3-diol
Catalog # APOSOR322236-100MG
Supplier:  Apollo Scientific
CAS Number:  
cis-4-Cyclopentene-1,3-diol
Catalog # APOSOR322236-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :100.116 g/mol
Storage Temperature:Ambient
CAS Number:29783-26-4
Formula:C₅H₈O₂
MDL Number:MFCD00077703