Order Entry
Czech Republic
ContactUsLinkComponent
3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR32216-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR32216-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.687 g/mol
Storage Temperature:Ambient
CAS Number:303986-05-2
Formula:C₁₁H₁₄ClNO₂
MDL Number:MFCD00793397