Order Entry
Czech Republic
ContactUsLinkComponent
(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile, tech
(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile, tech
Catalog # APOSOR32215-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile, tech
Catalog # APOSOR32215-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :269.342 g/mol
Storage Temperature:Ambient
CAS Number:303985-91-3
Formula:C₁₆H₁₉N₃O
MDL Number:MFCD00793385