Order Entry
Czech Republic
ContactUsLinkComponent
6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
Catalog # APOSOR32202-1G
Supplier:  Apollo Scientific
CAS Number:  
6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
Catalog # APOSOR32202-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.316 g/mol
Storage Temperature:Ambient
CAS Number:303146-59-0
Formula:C₁₅H₁₈N₂O
MDL Number:MFCD00793320