Order Entry
Czech Republic
ContactUsLinkComponent
4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one, tech
4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one, tech
Catalog # APOSOR32185-1G
Supplier:  Apollo Scientific
CAS Number:  
4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one, tech
Catalog # APOSOR32185-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.336 g/mol
Storage Temperature:Ambient
CAS Number:131365-81-6
Formula:C₁₇H₁₇N₃O
MDL Number:MFCD00140495