Order Entry
Czech Republic
ContactUsLinkComponent
4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
Catalog # APOSOR32183-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
Catalog # APOSOR32183-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.714 g/mol
Storage Temperature:Ambient
CAS Number:400075-09-4
Formula:C₁₅H₁₁ClN₂O
MDL Number:MFCD00232637