Order Entry
Czech Republic
ContactUsLinkComponent
3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32152-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32152-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.3 g/mol
Storage Temperature:Ambient
CAS Number:161185-41-7
Formula:C₁₅H₁₇NO₃
MDL Number:MFCD00101165