3-Acetyl-1-ethyl-4-hydroxy-1,2-dihydroquinolin-2-one 95%
Catalog # APOSOR32147-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.247 g/mol
Storage Temperature:Ambient
CAS Number:66134-57-4
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD00226605