Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1-Methylhydrazin-1-yl)pyrazine
2-(1-Methylhydrazin-1-yl)pyrazine 95%
2-(1-Methylhydrazin-1-yl)pyrazine 95%
2-(1-Methylhydrazin-1-yl)pyrazine 95%
APOSOR32144-1G
:
Apollo Scientific
:
2-(1-Methylhydrazin-1-yl)pyrazine 95%
APOSOR32144-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
124.144 g/mol
:
Ambient
:
76319-95-4
:
C₅H₈N₄
:
MFCD05975001