4-(1H-Pyrazol-4-yl)piperidine dihydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:690261-96-2
:
Formula:C₈H₁₃N₃·2ClH
MDL Number:MFCD17292786
:224.131 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together