
Methyl 3-iodopyrazine-2-carboxylate 97%
Catalog # APOSOR321370-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.021 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:173290-17-0
Formula:C₆H₅IN₂O₂
MDL Number:MFCD13195309