Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 97%
Irritant
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 97%
Catalog # APOSOR321366-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 97%
Catalog # APOSOR321366-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.193 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1245644-44-3
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD12756158