Order Entry
Czech Republic
ContactUsLinkComponent
1-(1H-Pyrazol-1-yl)cyclopropane-1-carboxylic acid 95%
Corrosive
Irritant
1-(1H-Pyrazol-1-yl)cyclopropane-1-carboxylic acid 95%
Catalog # APOSOR321344-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
1-(1H-Pyrazol-1-yl)cyclopropane-1-carboxylic acid 95%
Catalog # APOSOR321344-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :152.151 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:1548372-66-2
Formula:C₇H₈N₂O₂
MDL Number:MFCD28362645