
tert-Butyl (S)-3-(4-bromophenyl)piperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1476776-55-2
:
Formula:C₁₆H₂₂BrNO₂
MDL Number:MFCD27987933
:340.255 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together