
4,6-Dimethoxy-5-methyl-2,3-dihydro-1H-inden-1-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57122-06-2
:
Formula:C₁₂H₁₄O₃
MDL Number:MFCD28165860
:206.238 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together