Order Entry
Czech Republic
ContactUsLinkComponent
(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
Irritant
(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
Catalog # APOSOR321281-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
Catalog # APOSOR321281-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :263.198 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1821837-31-3
Formula:C₉H₁₄N₂S·BrH
MDL Number:MFCD30728434