Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate 95%
Irritant
tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate 95%
Catalog # APOSOR321276-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 4-(3-cyanophenyl)-3,6-dihydropyridine-1(2H)-carboxylate 95%
Catalog # APOSOR321276-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :284.353 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:370864-42-9
Formula:C₁₇H₂₀N₂O₂
MDL Number:MFCD24466376