Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 95%
Irritant
tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 95%
Catalog # APOSOR321275-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 4-(6-chloropyridin-3-yl)-3,6-dihydro-2H-pyridine-1-carboxylate 95%
Catalog # APOSOR321275-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :294.777 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1169699-77-7
Formula:C₁₅H₁₉ClN₂O₂
MDL Number:MFCD28978570