Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Irritant
Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Catalog # APOSOR321267-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl (S)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Catalog # APOSOR321267-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :269.767 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1388117-52-9
Formula:C₁₄H₁₉NO₂·ClH
MDL Number:MFCD30099275