Order Entry
Czech Republic
ContactUsLinkComponent
7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile, tech
7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile, tech
Catalog # APOSOR32125-1G
Supplier:  Apollo Scientific
CAS Number:  
7-(4-Chlorophenyl)-2-methyl-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile, tech
Catalog # APOSOR32125-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.701 g/mol
Storage Temperature:Ambient
CAS Number:860610-84-0
Formula:C₁₄H₉ClN₄
MDL Number:MFCD03787167