
2-Chloro-1',2',3',6'-tetrahydro-3,4'-bipyridine dihydrochloride 97%
Catalog # APOSOR321248-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:267.583 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Formula:C₁₀H₁₁ClN₂·2ClH
MDL Number:MFCD30099233