Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride 97%
Irritant
2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride 97%
Catalog # APOSOR321247-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methoxy-4-(piperidin-4-yl)pyridine dihydrochloride 97%
Catalog # APOSOR321247-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :265.179 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1137950-54-9
Formula:C₁₁H₁₆N₂O·2ClH
MDL Number:MFCD30099249