
4-(Piperidin-4-yl)phenol hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:263139-27-1
:
Formula:C₁₁H₁₅NO·ClH
MDL Number:MFCD10565007
:213.704 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together