Order Entry
Czech Republic
ContactUsLinkComponent
6-(Piperidin-4-yl)isoquinoline dihydrochloride 95%
Irritant
6-(Piperidin-4-yl)isoquinoline dihydrochloride 95%
Catalog # APOSOR321230-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(Piperidin-4-yl)isoquinoline dihydrochloride 95%
Catalog # APOSOR321230-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :285.212 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:2140866-96-0
Formula:C₁₄H₁₆N₂·2ClH
MDL Number:MFCD29075590