
5-(1,2,3,6-Tetrahydropyridin-4-yl)thiazole hydrochloride 97%
Catalog # APOSOR321216-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:202.704 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:2244084-18-0
Formula:C₈H₁₀N₂S·ClH
MDL Number:MFCD29075569