Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32109-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-(Dimethylamino)-1-(7-methoxy-1-benzofuran-2-yl)prop-2-en-1-one, tech
Catalog # APOSOR32109-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.274 g/mol
Storage Temperature:Ambient
CAS Number:477848-02-5
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD02082055