2-{[(4-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR32098-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.274 g/mol
Storage Temperature:Ambient
CAS Number:62370-45-0
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD01569055