2-{[(4-Bromophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR32097-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:294.144 g/mol
Storage Temperature:Ambient
CAS Number:341968-26-1
Formula:C₁₃H₁₂BrNO₂
MDL Number:MFCD01569054