2-{[(3,4-Dichlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR32094-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.138 g/mol
Storage Temperature:Ambient
CAS Number:341968-23-8
Formula:C₁₃H₁₁Cl₂NO₂
MDL Number:MFCD01569051