2-{[(4-Chlorophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR32092-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.693 g/mol
Storage Temperature:Ambient
CAS Number:62370-44-9
Formula:C₁₃H₁₂ClNO₂
MDL Number:MFCD00101155