Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one, tech
1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one, tech
Catalog # APOSOR32083-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Methylphenyl)-2-(1H-pyrazol-1-yl)ethan-1-one, tech
Catalog # APOSOR32083-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.236 g/mol
Storage Temperature:Ambient
CAS Number:321385-53-9
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD01315912