N'-[(1E)-1-(3-Methoxyphenyl)ethylidene]methoxycarbohydrazide, tech
Catalog # APOSOR32070-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.24 g/mol
Storage Temperature:Ambient
CAS Number:205121-88-6
Formula:C₁₁H₁₄N₂O₃
MDL Number:MFCD00792942