4-(4-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarbonitrile, tech
Catalog # APOSOR32066-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.729 g/mol
Storage Temperature:Ambient
CAS Number:67438-99-7
Formula:C₁₅H₁₂ClN₃
MDL Number:MFCD00663771