6-[(E)-2-Phenylethenyl]pyridazin-3-ol, tech
Catalog # APOSOR32062-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.221 g/mol
Storage Temperature:Ambient
CAS Number:83516-65-8
Formula:C₁₂H₁₀N₂O
MDL Number:MFCD00793247