Order Entry
Czech Republic
ContactUsLinkComponent
6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
Catalog # APOSOR32061-1G
Supplier:  Apollo Scientific
CAS Number:  
6-[(E)-2-Phenylethenyl]-2-(prop-2-en-1-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
Catalog # APOSOR32061-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.3 g/mol
Storage Temperature:Ambient
CAS Number:1421944-10-6
Formula:C₁₅H₁₆N₂O
MDL Number:MFCD00793235