Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Chloro-4-methoxyphenyl)-3-ethylurea
1-(3-Chloro-4-methoxyphenyl)-3-ethylurea, tech
1-(3-Chloro-4-methoxyphenyl)-3-ethylurea, tech
1-(3-Chloro-4-methoxyphenyl)-3-ethylurea, tech
APOSOR32049-1G
:
Apollo Scientific
:
1-(3-Chloro-4-methoxyphenyl)-3-ethylurea, tech
APOSOR32049-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
228.675 g/mol
:
Ambient
:
252058-85-8
:
C₁₀H₁₃ClN₂O₂
:
MFCD01316319